Would you like to log in again?
VWR® 11 mm Wide Opening Crimp-Top Vials
Supplier: VWR
VWR®
VW60825P-1232 VW60851T-1232 VW60871T-1232 VW60821W-1232 VW60782T-1232 VW60821G-1232
46610-740CS 520.36 USD InStock
46610-740 66009-850 46610-742 66009-870 66009-864 66010-539
VWR® 11 mm Wide Opening Crimp-Top Vials
Vials
Wide opening vials feature 40% larger neck opening than standard opening vials, for easier access and reduced needle damage.
  • 40% larger neck opening than standard vials
  • Easier access
  • Reduced needle damage
Vials are manufactured from 33 expansion borosilicate glass. Select amber vials for light-sensitive samples. Use 11 mm aluminum seals. Provided in clear-pack trays. The maximum fill volume is 1.8 ml.
Order Now
ORDER
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty