Would you like to log in again?
ACCUFORM® SSR™ Micro Vials, Borosilicate Glass, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
ACCUFORM®
60715-5 60715-2 60710-10 60710-2 60710-310 60710-110 60715-1 60710-3 60710-5 60715-310 60715-110 60710-1 60715-3
66010-621CS 249.36 USD InStock
66010-621 66010-632 66010-620 66010-631 66010-623 66010-622 66010-625 66010-624 66010-626 66010-629 66010-618 66010-628 66010-619
ACCUFORM® SSR™ Micro Vials, Borosilicate Glass, Kimble Chase, DWK Life Sciences
Vials
Storage and sample retrieval (SSR) vials are designed to provide reliable storage, retrieval, analysis, and delivery of liquid or powder samples.
These micro vials feature screw thread, open top closures, and a PTFE-faced silicone septum. These ungraduated vials are supplied assembled, and ready for use with autosamplers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty