Would you like to log in again?
Polypropylene Screw Caps, Linerless, Kimble Chase
Supplier: DWK Life Sciences
73805B-15415 73805B-18415 73805-15415 73805B-13415
66010-680CS 152.73 USD InStock
66010-680 66010-682 66010-678 89003-558
Polypropylene Screw Caps, Linerless, Kimble Chase
Closures Test/Sample Tube Closures
Polypropylene caps feature one-piece construction with a seal achieved by a unique interior design.
Caps withstand repeated autoclaving when loosened or removed from tubes.
Certifications: Designed from ASTM Specification E982
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty