Would you like to log in again?
Seals for 8 mm Crimp-Top Vials, Thermo Scientific
Supplier: Thermo Scientific
C4008-4A C4008-1A C4008-2A
66030-824CS 732.52 USD InStock
66030-824 66030-822 66011-180
Seals for 8 mm Crimp-Top Vials, Thermo Scientific
Closures Chromatography Vial Closures
National Scientific aluminum crimp closures provide a secure leak-resistant seal.
  • Aluminium crimp seals with prefitted septa
  • Pre-assembled caps and septa are convenient and minimize contamination from handling
Available with a variety of septa materials with and without liners to match your chromatography needs. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty