Would you like to log in again?
Dilution Vials with Cap, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
225002
66011-370CS 181.14 USD InStock
66011-370
Dilution Vials with Cap, WHEATON®, DWK Life Sciences
Vials
The vials are marked with a blue line at the 3 ml level. The vial comes with pre-attached solid-top black phenolic screw cap with 14B rubber liner.
Certifications: Dilution vials are manufactured from Wheaton33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty