Would you like to log in again?
OPTICLEAR™ Vials, Borosilicate Glass, with Polyethylene Stopper, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
OPTICLEAR®
60975L-5 60975L-4 60975L-1 60975L-3
66013-105CS 233.77 USD InStock
66013-105 66013-025 66013-069 66013-080
OPTICLEAR™ Vials, Borosilicate Glass, with Polyethylene Stopper, Kimble Chase, DWK Life Sciences
Vials
OPTICLEAR™ vials are supplied with polyethylene stoppers.
Vials have closed-bottom stoppers except for the 1dram size, which has open-bottom stoppers. Manufactured from N-51A borosilicate glass.
Ordering information: Vials come in lab pack cartons. Stoppers packed separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty