Would you like to log in again?
WHEATON® Polypropylene Vials with Insert, DWK Life Sciences
Supplier: DWK Life Sciences
225185
66014-265CS 502.88 USD InStock
66014-265
WHEATON® Polypropylene Vials with Insert, DWK Life Sciences
Vials
The 12×32 mm polypropylene vial with insert is an economical alternative to glass vials.
The conical interior of the vial ensures maximum retrieval of contents without the hassle of using removable inserts.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty