Would you like to log in again?
EPA Vials, Borosilicate Glass, with PTFE/Silicone-Lined Cap, Kimble Chase
Supplier: DWK Life Sciences
60961B-912 60961C-4 60961C-912 60961C-6
66014-304CS 1403.23 USD InStock
66014-304 66014-303 66014-295 66014-298
EPA Vials, Borosilicate Glass, with PTFE/Silicone-Lined Cap, Kimble Chase
Vials
For the collection and storage of water samples.
  • Made from KG-33 clear borosilicate glass
Clear glass vials have 0.005" PTFE-faced 0.120" silicone rubber septa with open-top screw-caps attached.
Comply with guidelines for establishing test procedures for the analysis of pollutants (Ref. EPA Methods 601, 602, 603, and 604).
Ordering information: Vials are packaged after the production process in shrink-wrapped modules to help reduce particulate contamination.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty