Would you like to log in again?
EPA Water Analysis Vials, Borosilicate Glass, Screw Thread, Without Closures, DWK Life Sciences
Supplier: DWK Life Sciences
60958A-11 60958A-6 60960A-912 60958A-16 60960A-4 60958A-912 60958A-4
66014-355CS 691.9 USD InStock
66014-355 66014-346 66014-323 66014-345 66014-301 66009-455 66014-353
EPA Water Analysis Vials, Borosilicate Glass, Screw Thread, Without Closures, DWK Life Sciences
Vials
Clear screw thread EPA vials for the collection and storage of water samples comply with the guidelines for establishing test procedures for the analysis of pollutants.
The EPA vials are fabricated from KG-33 borosilicate glass.
Ordering information: Open top closures and PFTE-faced silicone septa or solid top PFTE-faced white rubber-lined closures are available separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty