Would you like to log in again?
Open-Top Polypropylene Screw Thread Caps, Kimble Chase
Supplier: DWK Life Sciences
KIMBLE®
73806A-24400 73806A-20400 73806-24400 73806A-18400
66014-398CS 260.67 USD InStock
66014-398 66014-380 66009-967 66014-397
Open-Top Polypropylene Screw Thread Caps, Kimble Chase
Closures Test/Sample Tube Closures
Economical, one-piece construction caps are autoclavable and designed from ASTM Specification E982
Caps are open-top and do not come with a septa.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty