Would you like to log in again?
Lab File® Sample Vials, Borosilicate Glass, Wheaton, DWK Life Sciences
Supplier: DWK Life Sciences
Lab File®
224833 224802 224804 W224834 224805 224836 W224815 224832 224808 224801 224831 224806 224814
66011-813CS 142.86 USD InStock
66011-813 66011-824 66011-835 66011-857 66011-802 66011-846 66014-500 89500-052 66014-469 66014-436 66014-403 80076-576 66011-926
Lab File® Sample Vials, Borosilicate Glass, Wheaton, DWK Life Sciences
Vials
For storage of reagents and standards.
  • Excellent for small derivatization reactions
Wheaton 33® low extractable borosilicate glass which is ideal for research studies where stability of glass is of prime importance.
Certifications: Clear vials conform to USP Type I and ASTM E 438 Type I, Class A requirements. Amber vials made from borosilicate glass that conforms to USP Type I requirements for light transmission to protect light-sensitive products.
Ordering information: Without cap. For open-top caps, see 66012-598 series; for rubber-lined caps, see 66011-960 series.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty