Would you like to log in again?
Snap Caps, DWK Life Sciences
Supplier: DWK Life Sciences
242775 242772 242786 242776
89235-566CS 1063.86 USD InStock
89235-566 66014-612 89235-548 89235-556
Snap Caps, DWK Life Sciences
Closures Chromatography Vial Closures
Eliminate the need for crimping and decapping tools.
  • Manufactured from polyethylene
Fits the WHEATON E-Z vials® with snap ring top.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty