Would you like to log in again?
WHEATON® Caps for Screw Thread Vials, DWK Life Sciences
Supplier: DWK Life Sciences
242765
66014-734CS 185.95 USD InStock
66014-734
WHEATON® Caps for Screw Thread Vials, DWK Life Sciences
Closures Bottle Closures
Pre-assembled black polypropylene (PP) screw cap with septa.
  • With solid top
Suitable for 12×32 mm E-Z Vials™.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty