Would you like to log in again?
Sample Bottles, Wide Mouth, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
225534 225544 225543 225546 225538 225536 225532 225535
66015-010CS 298.58 USD InStock
66015-010 66015-020 66015-086 66015-064 66015-042 66015-224 66015-168 66015-166
Sample Bottles, Wide Mouth, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Sample bottles are designed for collecting, storing and exhibiting a wide variety of specimens in the lab or field.
  • Manufactured from Wheaton 800 soda-lime flint glass
PE caps snap over a special sealing ring to form an effective barrier to moisture, making these bottles especially suited for hygroscopic materials.
Packaging: Packed in convenient Lab File® cases.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty