Would you like to log in again?
VESSEL TEST DISSOLTN CLEAR 1L
Supplier: Thermo Scientific
Dissolution Test Vessels^ Manufactured from Type 1, class A borosilicate glass. Graduations in 50ml increments, 400ml to 1000ml. Meets all specifications of USP XXIV, Monograph\
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VESSEL TEST DISSOLTN CLEAR 1L 66016-166 100 002 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty