Would you like to log in again?
Scintillation Vials, Polypropylene, with Plug-Style Stopper, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
58552-4
66020-257CS 431.04 USD InStock
66020-257
Scintillation Vials, Polypropylene, with Plug-Style Stopper, Kimble Chase, DWK Life Sciences
Vials
Manufactured by injection molding from copolymer polypropylene to rigid specifications for low background characteristics, resulting in a container especially suitable for low-activity counting
  • Vials are seamless and have wide mouth openings
Plug-style stoppers permit fast closing and opening.
Packaging: Vials and caps are packed separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty