Would you like to log in again?
KIMBLE® ACCUFORM® Graduated High Recovery Vials, Borosilicate, DWK Life Sciences
Supplier: DWK Life Sciences
ACCUFORM®
60700-2 60700-1 60705-5 60700-3 60700-5 60705-3 60705-2 60700-10 60705-1
66020-904PK 344.02 USD InStock
66020-904 66020-905 66020-806 66020-902 66020-804 66020-903 66020-801 66020-802 66020-800
KIMBLE® ACCUFORM® Graduated High Recovery Vials, Borosilicate, DWK Life Sciences
Vials
These ACCUFORM® vials are internally contoured for optimal performance with limited samples.
  • Graduated to ensure more accurate measurements and dispensing
  • Tapered bases allow for maximum sample recovery
  • Open-top closure and PTFE-faced silicone septa, assembled and attached
Clear glass is manufactured from 33 expansion, low extractable borosilicate glass. Amber glass is manufactured from 51 expansion borosilicate amber glass.
Certifications: Clear glass conforming to USP Type I and ASTM E438, Type I, Class A requirements. Amber glass conforming to USP, Type I requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty