Would you like to log in again?
Omni-Vial® Vials, Plastic, Wheaton
Supplier: DWK Life Sciences
Omni-Vial®
225415 225414 225402
66021-250CS 677.64 USD InStock
66021-250 66021-249 66021-238
Omni-Vial® Vials, Plastic, Wheaton
Vials
Designed especially for beta and gamma counting, but can also be used for RIA, liquid scintillation counting, sample storage, sample counting, and chromatography.
  • Smooth interior allows easy transfer and access with a pipettor
  • The polyolefins used in these vials can be safely incinerated without the formation of hazardous gases
Available in polypropylene or high-density polyethylene. High-density polyethylene vials are available with a regular or hanging screw cap. The hanging cap features a lip that allows vial suspension and easy access when used with Wheaton glass and PET (polyethylene terephthalate) liquid scintillation vials (20 ml size with 22 mm neck size). The polypropylene vial is autoclavable.
Packaging: Vials and caps are packaged separately in polybags that contain 500 each. Also included is a polybag liner for easy disposal of the vials.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty