Would you like to log in again?
Scintillation Vials, Borosilicate Glass, with Screw Cap, Wheaton
Supplier: DWK Life Sciences
986580 986586 986541 986582 986540 986568 986560 986561 986542 986581 986548 986562 986546 986590 986591
66021-453CS 364.9 USD InStock
66021-453 66021-497 66021-675 66021-577 66021-533 66021-452 56941-386 66021-600 66021-645 66021-668 66021-657 66021-624 66021-646 66021-602 56941-513
Scintillation Vials, Borosilicate Glass, with Screw Cap, Wheaton
Vials
Made of low-potassium borosilicate glass to critical manufacturing tolerances, ensuring reliable, consistent results in scintillation counting.
  • Background counts remain low and ultraviolet transmission high
  • Vials are designed to load properly into scintillation counting equipment without jamming or breaking
Tight-fitting caps have tops suitable for writing. Vials with attached or unattached screw-caps.
Ordering information: Available with a variety of caps and liners. Packaged in individual cells in trays of 100.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty