Would you like to log in again?
Liquid Scintillation Vials, High-Density Polyethylene, with Screw Cap, Wheaton
Supplier: DWK Life Sciences
986721 986726 986711 986722 986724 986702 986706 986710 986720 986701 986714 986700 986704 986716
66021-695CS 523.7 USD InStock
66021-695 16300-228 16300-219 66021-680 66021-692 66021-704 16300-220 66021-715 66021-726 16300-222 66021-677 16300-224 66021-679 66021-690
Liquid Scintillation Vials, High-Density Polyethylene, with Screw Cap, Wheaton
Vials
High-density polyethylene walls increase counting accuracy.
  • Vials available as with caps attached or unattached
Made to precise tolerance for consistent results and efficient processing of samples.
Ordering information: Vials with caps attached are supplied in individual cells in trays of 100, 500 to a case. Vials with screw caps unattached are supplied 1000 vials per plastic bag with caps in separate bag.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty