Would you like to log in again?
White Urea Closures with Cork-Backed Aluminium Foil Liners, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
74520-15425 74520-22400
66022-140CS 435.68 USD InStock
66022-140 66022-150
White Urea Closures with Cork-Backed Aluminium Foil Liners, Kimble Chase, DWK Life Sciences
Closures Bottle Closures
Closure is made from white urea and features a cork-backed aluminum foil liner.
  • Top is suitable for marking
  • Ideal for scintillation vials
  • Closure is not autoclavable
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty