Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
WHEATON® Vial Racks, DWK Life Sciences
Supplier: DWK Life Sciences
868806 985750 868804 868805 868808 868810 985800
66022-580CS 388.53 USD InStock
66022-580 66022-525 66022-569 66022-478 66022-523 66022-556 14007-178
WHEATON® Vial Racks, DWK Life Sciences
Racks Cryogenic Vial Racks
Manufactured from polypropylene, vial racks are easy to clean and autoclavable
Each rack features alphanumeric indexing to simplify identification and selection. The racks are sturdy and stackable, even when vials are in place.
Ordering information: When selecting the proper rack, the vial O.D. cannot exceed the well I.D. dimension.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty