Would you like to log in again?
Nalgene® Unwire™ Racks, Polypropylene, Thermo Scientific
Supplier: Thermo Scientific
Nalgene® Unwire™
5976-0016 5976-0413 5976-0316 5976-0416 5976-0516 5976-0513 5976-0013 5976-0313
66023-540CS 230.29 USD InStock
66023-540 66023-530 66023-542 66023-520 66023-544 66023-534 66023-526 66023-528
Nalgene® Unwire™ Racks, Polypropylene, Thermo Scientific
Racks Tube Racks
These polypropylene racks are molded in one piece and have a three-tier, grid design
They withstand most chemicals and will not float or corrode in water baths. Letters and numbers on top tier aid in tube identification. Labels can be placed on large end plates. Racks for 13mm and 16mm tubes hold 72 tubes. Not autoclavable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty