Would you like to log in again?
Nalgene® Unwire™ Half-Racks, ResMer® Resin, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene® Unwire™
5972-0313 5972-0020 5972-0016 5972-0330 5972-0320 5972-0530 5972-0516 5972-0325 5972-0420 5972-0416 5972-0025 5972-0413 5972-0030 5972-0316 5972-0513 5972-0520 5972-0430 5972-0013
66024-018CS 230.15 USD InStock
66024-018 66024-016 66024-005 66023-980 66023-663 66024-002 66023-721 66023-787 66023-743 66024-010 66023-765 66024-000 66024-020 66023-889 66023-867 66023-845 66023-903 66023-969
Nalgene® Unwire™ Half-Racks, ResMer® Resin, Thermo Scientific
Racks Tube Racks
These racks are ideal for applications using 36 tubes or less
Racks for 13mm and 16mm tubes hold 36 tubes; racks for 20mm and 25mm tubes hold 20 tubes; racks for 30mm tubes hold 9 tubes.
These racks are made of polyketone.
They are molded of ResMer® resin, which has excellent chemical resistance and can be immersed in a water bath. Racks feature excellent thermal properties which allow both autoclaving and freezing. Letters and numbers on the racks aid in tube identification.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty