Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

11 mm Wide Opening Glass Crimp/Snap Top Vials, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

C4011-V1 C4011-6W C4011-9TR C4011-73W C4013-17 C4011-73P C4011-87AW C4011-9 C4011-V5 C4011-1Y C4011-S2W C4011-87 C4011-88W C4011-92 C4013-15P C4011-LV1 C4011-72W C4011-V6 C4011-87W C4011-1W C4011-LV2 C4011-S4 C4011-S5W C4011-73 C4011-89W C4011-S6W C4011-4 C4011-1 C4011-LV1W C4011-S1W C4011-88 C4011-1G C4011-1R C4011-1B C4011-S1 C4011-72P C4013-S15 C4011-95 C4011-5W C4011-2W C4011-LV2W C4011-6 C4011-5 C4011-74W C4011-S5
89235-962PK 78.34 USD InStock
89235-962 66064-230 66064-252 66064-210 89235-960 66064-254 66064-256 66064-212 66064-216 66030-550 66030-452 66030-556 66064-958 66011-241 66064-916 66030-554 66030-170 66011-181 66030-174 66030-130 66064-194 66064-250 66021-192 89235-950 66064-222 66064-246 89235-952 66064-248 66064-228 66064-920 66030-168 66030-124 66064-924 66030-128 66064-926 66030-126 66064-918 66065-260 66030-160 66030-120 69400-092 66030-162 66030-184 89235-904 66064-260
11 mm Wide Opening Glass Crimp/Snap Top Vials, Thermo Scientific
Vials
Use Thermo Scientific 11 mm Crimp/Snap Top Vials with snap caps or aluminum crimp seal closures. These high-quality 11 mm glass vials are 2 ml, 12×32 mm and fit most brands of autosamplers. Made of superior quality 33 expansion borosilicate clear Type 1, Class A or 51A Type 1, Class B amber glass, they meet all requirements of US, EU, and JPN Pharmacopia.

Easy-to-fill wide-neck opening vials require 6 mm diameter micro-inserts.

Save time during sample preparation and reduce the risk of contamination using Thermo Scientific 11 mm Crimp Top Clear Vial Convenience Kits (Unassembled). Each kit includes 100 clear glass vials and 100 crimp top caps with pre-assembled septa. Reusable two-compartment trays protect vials and closures, while keeping matching supplies together. Clear trays make it easy to keep track of available supplies without opening containers.

Identify sample and volume with graduated, write-on patch provided on certain vials. Get maximum sample extraction without the need for separate inserts by using Microsampling and High Recovery Vials. Obtain optimal recovery of critical polar, labile or OH-interacting compounds using silanized (deactivated) glass (optional). Polyspring inserts are self-aligning and provide a cushion against needle contact. Precision point inserts minimize residual sample loss. Pulled point inserts are an economical choice for noncritical applications. Compatible with most HPLC and GC autosamplers.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty