Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Inserts for National 13 mm Glass Crimp/Snap-Ring Vials

Supplier: Thermo Scientific
C4015-542 C4015-843 C4015-643 C4015-641 C4015-638
66065-380CS 344.58 USD InStock
66065-380 66011-202 66065-146 66030-396 66030-400
Inserts for National 13 mm Glass Crimp/Snap-Ring Vials
Vials
Thermo Scientific™ National vial inserts offer a variety of options incuding glass conical, polyspring and pulled point.

  • Have a nominal outer diameter of 6 or 8 mm to fit 15×45 (4 ml vials)
  • Deactivated inserts are NSC silanized

Autosampler compatibility includes Dionex, Shimadzu, Spark Holland, Varian, VWR (Merck)/Hitachi, and Waters (Wisp 48 Position Carousel).

Crimp/snap-ring finish for use with 13 mm aluminum seals or kim-snap closures.

Ordering information: Order caps separately.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty