Would you like to log in again?
Sample Vial Convenience Kits, Borosilicate Glass with Open-Top Screw Caps and Septa, Thermo Scientific
Supplier: Thermo Scientific
B7990-12 B7990-6
66030-704PK 264.33 USD InStock
66030-704 66030-698
Sample Vial Convenience Kits, Borosilicate Glass with Open-Top Screw Caps and Septa, Thermo Scientific
Vials
Unassembled convenience kits include clear, borosilicate glass vials with polypropylene, open-top screw caps, and natural PTFE/clear silicone septa.
Ordering information: Cartons contain shrink-wrapped vials and separately packaged caps and septa in plastic bags.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty