Would you like to log in again?
Target Glass Screw-Thread Vials, 10-425, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Target®
C4010-88A C4010-S2W CERT4010-91 C4010-S88 C4010-17A C4010-88AW C4010-2W C4010-88 C4010-2 C4010-1 C4010-67W C4010-88W C4010-1W C4010-17 C4010-17W C4010-S1W C4010-018
66030-308PK 178.16 USD InStock
66030-308 66030-922 66030-306 66030-916 66030-310 66065-682 66030-920 66030-312 69400-058 66030-258 66064-816 66064-808 66030-262 66030-260 66030-918 66064-180 66064-182
Target Glass Screw-Thread Vials, 10-425, Thermo Scientific
Vials
Vials made from Type I, Class A borosilicate glass.
  • 40% larger opening than standard vials
  • Silanized vials help to limit protein adsorption
  • Uniformly flat bottom for security with inserts
  • Ceramic write-on ID patch with graduations at 0.5, 1.0 and 1.5 ml
Inserts are designed for wide-opening 2 ml vials. Flat-bottom MicroSerts™ inserts are made from borosilicate glass. Conical-style Polyspring® inserts are available in borosilicate glass or polypropylene. Convenience kits include vials and caps with pre-inserted septa. Preassembled kits have vials with preattached caps and septa. Certified vial kits are tested and certified to meet or exceed up to 17 critical performance parameters; certificate enclosed with kit.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty