Would you like to log in again?
10-425 Wide Opening Glass Screw-Thread Vials, Thermo Scientific
Supplier: Thermo Scientific
C4010-57AW C4010-57W C4010-95 C4010-21 C4010-V1 C4010-57A C4010-017AW C4010-017W C4010-97W C4010-57 C4010-LV1 C4010-67A C4010-LV2 C4010-95W
43300-924PK 150.55 USD InStock
43300-924 66064-176 66064-872 66030-288 66033-060 66033-062 66064-868 66064-812 66033-064 66030-284 66030-274 66033-066 69400-050 66064-172
10-425 Wide Opening Glass Screw-Thread Vials, Thermo Scientific
Vials
These Thermo Scientific 10 mm wide opening screw-thread vials are manufactured from either clear Type 1, Class A glass or superior quality 51A amber Type 1, Class B glass.
Save time during sample preparation and reduce the risk of contamination with Thermo Scientific National 10 mm Assembled Convenience Kits. Kits include 100 amber glass vials assembled with 100 screw thread caps and septa. Kits are packaged in convenient vial trays with clear covers or in economical polybags.
Thermo Scientific National 10 mm Unassembled Convenience Kits save time during sample preparation and reduce the risk of contamination. Each kit includes 100 clear vials and 100 screw thread caps with pre-assembled septa. Reusable two-compartment trays protect vials and closures, while keeping matching supplies together. Clear trays make it easy to keep track of available supplies without opening containers.
Vials have a 10-425 thread finish. Get maximum sample extraction without the need for separate inserts by using Microsampling and High Recovery Vials. Identify sample and volume using the graduated, write-on patch provided on certain vials. Obtain maximum recovery of critical polar, labile or OH-interacting compounds using silanized (deactivated) glass (optional). Polyspring inserts are self-aligning and provide a cushion against needle contact. Precision point inserts minimize residual sample loss. Autosampler compatibility includes Jasco, PerkinElmer, Shimadzu, Varian, and Waters. See the Thermo Scientific Columns and Consumables Catalog for autosampler compatibility.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty