Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
National™ Micro-Centrifugal Filters, Nonsterile, Thermo Scientific
Supplier: Thermo Scientific
F2517-5 F2517-3 F2517-10 F2517-2 F2517-6 F2517-1 F2517-4 F2517-8 F2517-7 F2517-9
66064-450PK 394.26 USD InStock
66064-450 66064-442 66064-452 66064-444 66064-454 66064-446 66064-456 66064-448 97006-922 97006-920
National™ Micro-Centrifugal Filters, Nonsterile, Thermo Scientific
Filters Centrifugal Filters
Filter volumes as low as 50 µl with low hold-up volume with these National™ micro-centrifugal filters.
  • 10000×g maximum centrifugal force
  • Non sterile
  • Bulk packed
The filters feature a virgin polypropylene filter housing with tapered 2 ml, capped receiver tube, and can be used with any benchtop microcentrifuge.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty