Would you like to log in again?
National™ Centrifugal Filters, Non sterile,25 ml, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
F2519-2 F2519-1 F2519-4 F2519-5 F2519-3 F2519-6
66064-462PK 394.99 USD InStock
66064-462 66064-464 66064-466 66064-468 66064-458 66064-460
National™ Centrifugal Filters, Non sterile,25 ml, Thermo Scientific
Filters Centrifugal Filters
Filter volumes up to 25 ml with these National™ 25 ml centrifugal filters.
  • 2500×g maximum centrifugal force
  • Non sterile
  • Bulk packed
The filters feature a virgin polypropylene filter housing with conical receiver, and can be used with any benchtop or floor model laboratory centrifuge.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty