Would you like to log in again?
National™ Centrifugal Filters, Non sterile, 2 ml, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
F2520-3 F2520-5 F2520-7 F2520-8 F2520-1 F2520-2 F2520-4 F2520-6
66064-484PK 238.48 USD InStock
66064-484 66064-472 66064-474 66064-476 66064-478 66064-480 66064-482 66064-470
National™ Centrifugal Filters, Non sterile, 2 ml, Thermo Scientific
Filters Centrifugal Filters
Filter volumes up to 2 ml with these National™ centrifugal filters.
  • 5000×g maximum centrifugal force
  • Non sterile
  • Bulk packed
Filters feature a virgin polypropylene filter housing with capped receiver tube, and can be used with any benchtop or floor model laboratory centrifuge.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty