Would you like to log in again?
13-425 Screw-Thread Vials
Supplier: Thermo Scientific
C4015-017A CERT4015-86 C4015-93 C4015-14 C4015-S11W C4015-9 C4015-21AW C4015-2W C4015-S1 C4015-S2W C4015-S15 C4015-17W C4015-11W C4015-88 C4015-482A C4015-21W C4015-017 C4015-2 C4015-1 C4015-88AW C4015-21 C4015-S2 C4015-17 C4015-17AW C4015-17A
66064-330PK 134.08 USD InStock
66064-330 66030-516 66064-332 66064-334 66065-138 66030-498 66065-674 66030-510 66030-478 69400-080 66030-494 66030-492 66030-484 66030-386 66064-328 66065-222 69400-078 66065-142 66030-488 66030-380 66030-482 66030-384 66030-480 66033-022 66030-382
13-425 Screw-Thread Vials
Vials
These 4 ml, 15×45 mm vials feature a 13-425 thread finish.
Thermo Scientific National unassembled 4 ml screw thread vial convenience kits include 100 vials and 100 caps with pre-assembled septa.
These vials are made of borosilicate glass material.
All items remain handy and dust-free. Caps with pre-inserted septa are not attached to vials; packaged in polybags.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty