Would you like to log in again?
Storage Vials and Closures
Supplier: Thermo Scientific
B7800-4 B7990-2A B7800-1-9A B7800-20A B7990-5 B7800-5 B7800-3A B7990-4 B7800-6A B7800-2A B7800-1A B7800-6 B7990-12A B7800-12A B7800-1-9 B7990-2 B7990-1 B7990-6A B7800-4A B7800-20 B7990-3 B7990-1A
66030-712PK 332.9 USD InStock
66030-712 66030-702 76414-876 66030-708 66065-602 76414-874 66030-716 66030-706 75839-124 66030-684 66030-674 66030-696 66030-682 66030-694 66030-688 66030-700 66030-678 66030-686 66030-676 66030-680 66030-692 66030-690
Storage Vials and Closures
Vials
These storage screw thread vials are manufactured from superior quality 33 expansion borosilicate clear Type 1, Class A or 51A amber Type 1, Class B glass.
  • PTFE-lined solid-top storage caps
  • Not autoclavable
  • PTFE on film/foam packing offers broad chemical resistance
The storage vials range from 2 to 40 ml.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty