Would you like to log in again?
Silanized Sample Vial
Supplier: Thermo Scientific
B7999-S12A
66065-612PK 246.27 USD InStock
66065-612
Silanized Sample Vial
Vials
Improve sample analysis sensitivity by eliminating sample/glass reactions.
  • Superior quality borosilicate clear or amber glass
  • Eliminate leaching of ions
  • Provide consistent pH for duration of sample storage life
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty