Would you like to log in again?
SP Bel-Art Wedge Pan, Bel-Art Products, a part of SP
Supplier: Bel-Art
F24894-0000
66104-100CS 599.5 USD InStock
66104-100
SP Bel-Art Wedge Pan, Bel-Art Products, a part of SP
Dustpans
Great for chemical spill clean-up
  • Inexpensive
  • Disposable
  • Convenient to use
Thin-edged wedge pan scoops up the refuse for insertion into the proper disposal receptacle. A single pan is useful in most instances. For large spills, two pans pushed toward one another will gather the spill more quickly. Pan measures 7x71/4" and tapers from a thin edge to 11/4" deep. The recessed hand grip provides room for fingers and thumb.
Use to scoop up spilled chemicals after containment by an absorbent medium.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty