Would you like to log in again?
Bytac® Teflon® Resin Surface Protectors, Saint-Gobain Performance Plastics
Supplier: Saint-Gobain
Bytac®
D9860003 D1069324 D9860011 D9860005 D1069329 D9860008 D9860002 D9860012
66110-950EA 269.44 USD InStock
66110-950 54112-090 54112-256 66110-958 66110-956 54112-100 66110-954 54112-085
Bytac® Teflon® Resin Surface Protectors, Saint-Gobain Performance Plastics
Surface Protectors Bench Protectors
Put the benefits of Teflon® resin on any flat surface with this pressure-sensitive overlay
Available with two different FEP bonded support sheets. Aluminum-backed overlay can withstand temperatures up to 205°C (400°F). Vinyl-backed overlay can withstand temperatures up to 93°C (200°F).
Create a non-stick, easy-to-clean, easy-to-apply work surface. Non-contaminating and chemically inert. Ideal as bench-top covering or release liner for inside surfaces. Overlay is made of 0.02mm (0.001") thick Teflon® resin. Easily cut with scissors.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty