Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Versilic® Peroxide Cured Silicone Stoppers, Saint-Gobain

Supplier: Saint-Gobain

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Versilic®
DX263013-50 DX263006-50 DX263107-01 DX263075-02 DX263045-05 DX263034-05 DX263054-02 DX263008-50 DX263021-20 DX263070-02 DX263092-01 DX263100-01 DX263038-05 DX263015-50
66111-270PK 54.68 USD InStock
66111-270 66111-292 66111-280 66111-276 66111-254 66111-286 66111-288 66111-294 66111-272 66111-260 66111-252 66111-296 66111-246 66111-248
Versilic® Peroxide Cured Silicone Stoppers, Saint-Gobain
Stoppers Silicone Stoppers
Highly recommended for all types of tissue culture work.

  • Non-toxic
  • Chemically and biologically neutral
  • Temperature up to 250 °C (482 °F)

They are suitable for use at temperatures between –70 to 250 °C (–94 to 482 °F).

These stoppers are odorless, tasteless, non-adherent and resistant to aging.

They are ideal in biology, microbiology and hematology, as well as food science, technical, medical and hospital apparatus.

Certifications: DIN Standard 18271, FDA/BGA approved.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty