Would you like to log in again?
CHEMFLUOR® Fluoropolymer Films, PTFE, Saint Gobain
Supplier: Saint-Gobain
D1069279 D1069278
66111-550EA 319.8 USD InStock
66111-550 66111-552
CHEMFLUOR® Fluoropolymer Films, PTFE, Saint Gobain
Surface Protectors Bench Protectors
These premium grade, reusable, and easy-to-clean skived films are made from virgin PTFE
  • Temperature and corrosion resistant
  • Non-stick surface
  • Non-porous and non-absorbent
The film will not stain or retain odors, flavors or other contaminants. Cut to size and use as weighing sheets.
The non-stick surface, temperature, and corrosion resistance features make Chemfluor film an ideal lining or covering for shelves, drawers, tables, trays, drain boards, and sinks.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty