Would you like to log in again?
Chemware® PFA Test Tubes, Saint-Gobain Performance Plastics
Supplier: Saint-Gobain
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Chemware®
D1069695 D1069599 D1069598
66111-564EA 112.85 USD InStock
66111-564 66111-574 66111-578
Chemware® PFA Test Tubes, Saint-Gobain Performance Plastics
Tubes Test Tubes
These unbreakable test tubes are excellent for trace metal analysis.
  • Made from fluoropolymer PFA resin
  • Low metal ion content
  • Inert
They are non-contaminating, and transparent for easy viewing of contents. They withstand extreme temperature. Can be sterilized repeatedly.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty