Would you like to log in again?
AMCO Clear® Primary Turbidity Standards, GFS Chemicals
Supplier: GFS Chemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AMCO Clear®
85043 85050 86110 85075 85047 85069 85159 85009 85141 85151 85040 85166 85092 85093 85142 85160 85122 85123 85027 85005 85150 85014 85030 85056 85164 85016 85062 85068 85051 85031 85026 85007 85063 85024 85034 85011 85018 85074
66115-016EA 130.91 USD InStock
66115-016 66115-038 66115-136 66115-018 66115-138 66115-034 66115-154 66115-132 66115-014 66115-156 66115-134 66115-150 66115-172 66115-010 66115-152 66115-174 66115-130 66115-170 66115-008 66115-148 66115-026 66115-004 66115-128 66115-028 66115-006 66115-166 66115-144 66115-022 66115-000 66115-146 66115-046 66115-002 66115-024 66115-162 66115-140 66115-142 66115-042 66115-020
AMCO Clear® Primary Turbidity Standards, GFS Chemicals
Standards Turbidity Standards
EPA-approved primary turbidity standards for calibrating turbidimeters and nephelometers
One year shelf life.
Ready to use at their stated value without mixing or diluting. Composed of submicron solid polymer beads suspended in ultra-pure water. Guaranteed accuracy within 1%. Safe, non-toxic, non-carcinogenic alternative to Formazin. Available for non-ratio and ratio meters.
Certifications: Approved by EPA, Standard Methods, ASTM, ASBC, and EBC. Traceable to NIST 1690, 1691.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty