Would you like to log in again?
Pops® DPD Free Chlorine Reagent Powder, HF Scientific
Supplier: HF Scientific
10312 10300 10307 10301 10306
66130-400EA 313.56 USD InStock
66130-400 66130-410 66130-402 66130-416 66130-408
Pops® DPD Free Chlorine Reagent Powder, HF Scientific
Environmental Test Kits
For determination of free chlorine by the DPD method
This individually packaged doses of reagent used to deliver a measured amount of DPD Reagent to a 5, 10 or 25 mL testing sample.
Ordering information: Pack of 200 or 1000 units.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty