Would you like to log in again?
Total Iron LRC Comparator Kit, LaMotte
Supplier: LaMotte
7787-01
66170-284EA 231.16 USD InStock
66170-284
Total Iron LRC Comparator Kit, LaMotte
Electrical Meters Voltmeters
This kit tests for Low Iron in water using Iron Reagents and the LRC Comparator
Range/Sensitivity is 0.05, 0.10, 0.20, 0.30, 0.40, 0.60, 0.80, 1.0 ppm Fe.
Delivery information: It includes 30 mL Iron Reagent #1, 4.5 g Iron Reagent #2 powder, 1 to 5 mL distilled water ampoule, 3 glass test tubes of 10 mL with caps, 1 plastic spoon 0.15 g and Low Range Comparator.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty