Would you like to log in again?
Sub Surface Sampler, Grab Sampler® I, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Grab Sampler®
990250
66175-071EA 1156.94 USD InStock
66175-071
Sub Surface Sampler, Grab Sampler® I, DWK Life Sciences
Samplers Fluid Samplers
Ideal for sampling from spillways, docks and other sub surfaces.
  • Lightweight construction
  • Eliminates surface contamination
  • No need to physically enter the sample area
  • Helps meet EPA requirements for water and wastewater sampling
The basic unit features a 183 cm aluminum body with clamp for securing a 1000 ml sample bottle. An attached extension allows the bottle to be opened and closed under water for discrete sampling.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty