Would you like to log in again?
Norwell® Economy Bailers, Saint Gobain Performance Plastics
Supplier: Saint-Gobain
D8653075
66175-296EA 225.27 USD InStock
66175-296
Norwell® Economy Bailers, Saint Gobain Performance Plastics
Samplers Fluid Samplers
Clear, non-graduated body made of FEP resin and a secure handle to provide a convenient and secure method of sampling water from a well or container.
  • Reusable and economical
  • Chemically resistant PTFE and FEP resin fabrication
  • Graduation marks and transparent body for sample monitoring
  • PTFE ground ball check valve secures fluids in bailer
  • Easy to disassemble and clean
Features a PTFE ground ball check valve that allows water to enter the cylinder and be retained when the bailer is raised. The top and bottom are constructed of PTFE fluoropolymer, and easily disassemble for cleaning. Fits into wells or containers that exceed 2”.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty