Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Nalgene Magnetic Carboy Stirrer, Thermo Scientific
Supplier: Thermo Scientific
DS2227-0020
66186-041CS 731.68 USD InStock
66186-041
Nalgene Magnetic Carboy Stirrer, Thermo Scientific
Stirrers Magnetic Stirrers
Nalgene Magnetic Carboy Stirrer is used with Nalgene 10- and 20L carboys for low-speed mixing of high volumes of media and buffer solution
The stirrer adjusts easily to fit either a 10 or 20L Nalgene carboy with an 83B size closure. The magnetic stirrer package includes closure and two impellers. Stir bar is designed as a 13mm diameter and shaft O.D: 13mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty