Would you like to log in again?
Sugar Refractometer, 28 to 62%, Sper Scientific
Supplier: Sper Scientific
300002
66214-976EA 298.4 USD InStock
66214-976
Sugar Refractometer, 28 to 62%, Sper Scientific
Refractometers Handheld Refractometers
Designed to test solutions with a higher sugar content, such as fruit juice concentrates, canned fruits containing added sugar, or ketchup
The unit will also test for egg yolk. To operate, simply place a drop or two of sample onto the prism and read the results. Supplied with a hard carrying case and calibration tool.
Ordering information: Refractometer is supplied with a 5-year warranty.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty