Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rocker II™ Platform Mixer, 230 V

Supplier: Boekel Scientific
Rocker II™
260350-2
66238-334EA 1369.06 USD InStock
66238-334
Rocker II™ Platform Mixer, 230 V
Shakers and Mixers Rotating/Rocking Shakers
Front to back rocking motion is ideal for gel staining and destaining, Northern, Western, and Southern blots, mixing test tubes, and rocking agglutination cards.

  • 230 V model
  • Tilt angle is adjustable from 0 to 20°
  • Speed is adjustable from 3 to 25 strokes per minute
  • Both tilt angle and speed are adjustable without tools

Rocker accepts a wide variety of flasks, plates, and tubes. Thin power cord easily passes through incubator door gaskets.

Certifications: The 230 V model is CE marked.

Ordering information: Supplied with flat, non-skid mat. Low voltage (12 V) power is supplied by wall mount transformer (included). Accessories sold separately.

Caution: 230 V model. Corresponding model with voltage suitable for use in US (66238-336) is also available, please see separate product page.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty