Would you like to log in again?
BREATHING TUBE ASSY 26
Supplier: 3M OCCUPATIONAL HEALTH
Breathing Tube Assembly^ This assembly, 26 inches long, is used with the 3M* Breathe Easy* 10 Powered Air Purifying(PAPR) System. It can be used with the Tychem* QC hood or the butyl rubber hood system.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BREATHING TUBE ASSY 26 67100-268 7000148506 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty