Would you like to log in again?
BREATHING TUBE
Supplier: 3M OCCUPATIONAL HEALTH
Breathing Tube^ Part of the 3M* Air Regulating Valve Assembly W-3061(67100-816) which allows the 3M* Full Facepiece 7800S or the 3M* Full Facepieces 6000DIN to be operated in airline mode. The breathing tube connects to the 3M* Airline Adapter W-3187 (67101-306) in the center part of the facepiece.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BREATHING TUBE 67100-888 7010319407 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty